Tisk…

7-Hydroxy-3,4-dihydro-2H-isoquinolin-1-one 95%

Dodavatel: Apollo Scientific

OR925654-100MG OR925654-1G OR925654-250MG
APOSOR925654-100MGEA 2030 CZK
APOSOR925654-100MG APOSOR925654-1G APOSOR925654-250MG
7-Hydroxy-3,4-dihydro-2H-isoquinolin-1-one 95%
7-Hydroxy-3,4-dihydro-2H-isoquinolin-1-one
Vzorec: C₉H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD04114872
CAS číslo: 22246-05-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR