3-Methyl-1,5,6,7-tetrahydroindazol-4-one 95%
Dodavatel: Apollo Scientific
OR925641-1G
OR925641-250MG
APOSOR925641-1GEA
13660
CZK
APOSOR925641-1G
APOSOR925641-250MG
3-Methyl-1,5,6,7-tetrahydroindazol-4-one 95%
3-Methyl-1,5,6,7-tetrahydroindazol-4-one
Vzorec:
C₈H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00168632 CAS číslo: 63446-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...