Tisk…
Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
OR925621-5G
APOSOR925621-5GEA 3190 CZK
APOSOR925621-5G
Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate 95%
Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
Vzorec: C₇H₁₀N₂O₂
MW: 154,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204177
CAS číslo: 10250-59-6