Tisk…

5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester 95%

Dodavatel: Apollo Scientific

OR925600-1G OR925600-250MG
APOSOR925600-1GEA 20470 CZK
APOSOR925600-1G APOSOR925600-250MG
5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester 95%
5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester
Vzorec: C₇H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD24843138
CAS číslo: 80196-63-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR