Tisk…

(S)-Methyl pyrrolidine-3-carboxylate hydrochloride 98%

Dodavatel: Apollo Scientific

OR925582-1G OR925582-5G
APOSOR925582-1GEA 2940 CZK
APOSOR925582-1G APOSOR925582-5G
(S)-Methyl pyrrolidine-3-carboxylate hydrochloride 98%
(S)-Methyl pyrrolidine-3-carboxylate hydrochloride
Vzorec: C₆H₁₁NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD11840196
CAS číslo: 1099646-61-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR