3-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR925560-10G
OR925560-1G
OR925560-5G
APOSOR925560-10GEA
14290
CZK
APOSOR925560-10G
APOSOR925560-1G
APOSOR925560-5G
3-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 95%
3-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Vzorec:
C₁₁H₁₀N₂O MW: 186,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02705915 CAS číslo: 21487-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...