Tisk…

4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%

Dodavatel: Apollo Scientific

OR925546-1G OR925546-5G
APOSOR925546-1GEA 6220 CZK
APOSOR925546-1G APOSOR925546-5G
4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester 95%
4-Amino-3-methyl-pyrazole-1-carboxylic acid tert-butyl ester
Vzorec: C₉H₁₅N₃O₂
MW: 197,234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23115361
CAS číslo: 847139-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR