Tisk…

4-(4-Chloro-phenyl)-isoxazol-5-ylamine 95%

Dodavatel: Apollo Scientific

OR925457-1G OR925457-250MG
APOSOR925457-1GEA 9530 CZK
APOSOR925457-1G APOSOR925457-250MG
4-(4-Chloro-phenyl)-isoxazol-5-ylamine 95%
4-(4-Chloro-phenyl)-isoxazol-5-ylamine
Vzorec: C₉H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD01747260
CAS číslo: 64047-49-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR