2-(Butylthio)pyrimidine-5-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR925443-1G
OR925443-5G
APOSOR925443-1GEA
4770
CZK
APOSOR925443-1G
APOSOR925443-5G
2-(Butylthio)pyrimidine-5-carbaldehyde 95%
2-(Butylthio)pyrimidine-5-carbaldehyde
Vzorec:
C₉H₁₂N₂OS MW: 196,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08691652 CAS číslo: 915920-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...