7-Nitro-imidazo[1,2-a]pyridine 95%
Dodavatel: Apollo Scientific
OR924836-1G
OR924836-250MG
APOSOR924836-1GEA
23580
CZK
APOSOR924836-1G
APOSOR924836-250MG
7-Nitro-imidazo[1,2-a]pyridine 95%
7-Nitro-imidazo[1,2-a]pyridine
Vzorec:
C₇H₅N₃O₂ MW: 163,134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17016107 CAS číslo: 1234615-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...