Tisk…

3-(Methoxymethyl)cyclobutan-1-amine

Dodavatel: Apollo Scientific
Varování

OR924769-1G OR924769-5G
APOSOR924769-1GEA 17360 CZK
APOSOR924769-1G APOSOR924769-5G
3-(Methoxymethyl)cyclobutan-1-amine
3-(Methoxymethyl)cyclobutan-1-amine
Vzorec: C₆H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD15474858
CAS číslo: 1209654-41-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR