(3S)-3-Phenylpyrrolidine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR924732-1G
OR924732-250MG
APOSOR924732-1GEA
25790
CZK
APOSOR924732-1G
APOSOR924732-250MG
(3S)-3-Phenylpyrrolidine 98%
(3S)-3-Phenylpyrrolidine
Vzorec:
C₁₀H₁₃N MW: 147,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19216484 CAS číslo: 62624-46-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...