1-(3-Iodobenzyl)-1H-1,2,4-triazole 95%
Dodavatel: Apollo Scientific
OR9246-1G
OR9246-250MG
APOSOR9246-1GEA
7670
CZK
APOSOR9246-1G
APOSOR9246-250MG
1-(3-Iodobenzyl)-1H-1,2,4-triazole 95%
1-(3-Iodobenzyl)-1H-1,2,4-triazole
Vzorec:
C₉H₈IN₃ Storage Temperature: Ambient |
MDL Number:
MFCD08271930 CAS číslo: 615534-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...