Tisk…

Methyl 5-methyloxazole-2-carboxylate 95%

Dodavatel: Apollo Scientific

OR924492-1G OR924492-250MG
APOSOR924492-1GEA 22950 CZK
APOSOR924492-1G APOSOR924492-250MG
Methyl 5-methyloxazole-2-carboxylate 95%
Methyl 5-methyloxazole-2-carboxylate
Vzorec: C₆H₇NO₃
MW: 141,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839097
CAS číslo: 124999-43-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR