Tisk…

N,N-Dimethyl-2-phenylacetamide 95%

Dodavatel: Apollo Scientific

OR924304-1G OR924304-250MG
APOSOR924304-1GEA 4040 CZK
APOSOR924304-1G APOSOR924304-250MG
N,N-Dimethyl-2-phenylacetamide 95%
N,N-Dimethyl-2-phenylacetamide
Vzorec: C₁₀H₁₃NO
MW: 163,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00157672
CAS číslo: 18925-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR