Tisk…

2-Bromo-6-methylnicotinonitrile 95%

Dodavatel: Apollo Scientific

OR924227-1G OR924227-250MG
APOSOR924227-1GEA 3710 CZK
APOSOR924227-1G APOSOR924227-250MG
2-Bromo-6-methylnicotinonitrile 95%
2-Bromo-6-methylnicotinonitrile
Vzorec: C₇H₅BrN₂
MW: 197,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254423
CAS číslo: 155265-57-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR