1-Butyl-2-chloro-1H-imidazole 95%
Dodavatel: Apollo Scientific
OR924177-1G
OR924177-250MG
APOSOR924177-1GEA
14350
CZK
APOSOR924177-1G
APOSOR924177-250MG
1-Butyl-2-chloro-1H-imidazole 95%
1-Butyl-2-chloro-1H-imidazole
Vzorec:
C₇H₁₁ClN₂ MW: 158,629 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10699141 CAS číslo: 1053655-55-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...