Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
OR924117-1G
OR924117-250MG
APOSOR924117-1GEA
5790
CZK
APOSOR924117-1G
APOSOR924117-250MG
Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate
Vzorec:
C₇H₉ClN₂O₂ MW: 188,612 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19704132 CAS číslo: 173841-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...