(S)-Fmoc-2-amino-5-(trityl-carbamoyl)pentanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR924095-100MG
OR924095-250MG
APOSOR924095-100MGEA
2240
CZK
APOSOR924095-100MG
APOSOR924095-250MG
(S)-Fmoc-2-amino-5-(trityl-carbamoyl)pentanoic acid
(S)-Fmoc-2-amino-5-(trityl-carbamoyl)pentanoic acid
Vzorec:
C₄₀H₃₆N₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD03093573 CAS číslo: 1263046-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...