4-Methoxy-3,5-dimethylpicolinic acid 95%
Dodavatel: Apollo Scientific
OR924043-1G
OR924043-250MG
APOSOR924043-1GEA
5920
CZK
APOSOR924043-1G
APOSOR924043-250MG
4-Methoxy-3,5-dimethylpicolinic acid 95%
4-Methoxy-3,5-dimethylpicolinic acid
Vzorec:
C₉H₁₁NO₃ MW: 181,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22397804 CAS číslo: 138569-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...