Tisk…

3-Chloro-2-(hydroxymethyl)thiophene 95%

Dodavatel: Apollo Scientific

OR924020-1G OR924020-5G
APOSOR924020-1GEA 8270 CZK
APOSOR924020-1G APOSOR924020-5G
3-Chloro-2-(hydroxymethyl)thiophene 95%
3-Chloro-2-(hydroxymethyl)thiophene
Vzorec: C₅H₅ClOS
MW: 148,611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203004
CAS číslo: 193602-41-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR