2,2-Dimethyl-1H-indene-1,3(2H)-dione 95%
Dodavatel: Apollo Scientific
OR923993-1G
OR923993-250MG
APOSOR923993-1GEA
8830
CZK
APOSOR923993-1G
APOSOR923993-250MG
2,2-Dimethyl-1H-indene-1,3(2H)-dione 95%
2,2-Dimethyl-1H-indene-1,3(2H)-dione
Vzorec:
C₁₁H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00144912 CAS číslo: 17190-77-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...