Tisk…

6-Aminonicotinaldehyde 95%

Dodavatel: Apollo Scientific

OR923967-1G OR923967-5G
APOSOR923967-1GEA 2690 CZK
APOSOR923967-1G APOSOR923967-5G
6-Aminonicotinaldehyde 95%
6-Aminonicotinaldehyde
Vzorec: C₆H₆N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09757481
CAS číslo: 69879-22-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR