(R)-2-Ethyl-piperazine 95%
Dodavatel: Apollo Scientific
OR923836-1G
OR923836-250MG
APOSOR923836-1GEA
21490
CZK
APOSOR923836-1G
APOSOR923836-250MG
(R)-2-Ethyl-piperazine 95%
(R)-2-Ethyl-piperazine
Vzorec:
C₆H₁₄N₂ MW: 114,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411921 CAS číslo: 393781-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...