Tisk…

1-Benzyl-1,2-dihydropyridin-2-one 95%

Dodavatel: Apollo Scientific

OR923763-1G OR923763-5G
APOSOR923763-1GEA 9330 CZK
APOSOR923763-1G APOSOR923763-5G
1-Benzyl-1,2-dihydropyridin-2-one 95%
1-Benzyl-1,2-dihydropyridin-2-one
Vzorec: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00462400
CAS číslo: 1753-62-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR