Tisk…

(S)-5-Phenylmorpholin-2-one 95%

Dodavatel: Apollo Scientific

OR923728-1G OR923728-5G
APOSOR923728-1GEA 2480 CZK
APOSOR923728-1G APOSOR923728-5G
(S)-5-Phenylmorpholin-2-one 95%
(S)-5-Phenylmorpholin-2-one
Vzorec: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269693
CAS číslo: 144896-92-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR