Tisk…

Ethyl 1-methyl-2-oxopiperidine-3-carboxylate 95%

Dodavatel: Apollo Scientific

OR923597-1G OR923597-250MG
APOSOR923597-1GEA 20470 CZK
APOSOR923597-1G APOSOR923597-250MG
Ethyl 1-methyl-2-oxopiperidine-3-carboxylate 95%
Ethyl 1-methyl-2-oxopiperidine-3-carboxylate
Vzorec: C₉H₁₅NO₃
MW: 185,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11106791
CAS číslo: 21576-27-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR