Tisk…

Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride 95%

Dodavatel: Apollo Scientific

OR923595-1G OR923595-250MG
APOSOR923595-1GEA 5360 CZK
APOSOR923595-1G APOSOR923595-250MG
Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride 95%
Ethyl 2-(pyrrolidin-3-yl)acetate hydrochloride
Vzorec: C₈H₁₅NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD17171210
CAS číslo: 726139-60-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR