2-Bromo-5-chloropyrimidine 95%
Dodavatel: Apollo Scientific
OR923484-1G
OR923484-250MG
APOSOR923484-1GEA
1490
CZK
APOSOR923484-1G
APOSOR923484-250MG
2-Bromo-5-chloropyrimidine 95%
2-Bromo-5-chloropyrimidine
Vzorec:
C₄H₂BrClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD12026299 CAS číslo: 124405-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...