8-Oxa-3-aza-bicyclo[3.2.1]octane 98%
Dodavatel: Apollo Scientific
Synonyms:
8-Oxa-3-azabicyclo[3.2.1]octane
OR923369-1G
OR923369-250MG
APOSOR923369-1GEA
8900
CZK
APOSOR923369-1G
APOSOR923369-250MG
8-Oxa-3-aza-bicyclo[3.2.1]octane 98%
8-Oxa-3-aza-bicyclo[3.2.1]octane
Vzorec:
C₆H₁₁NO MW: 113,158 g/mol Bod varu: 176…178 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09836264 CAS číslo: 280-13-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...