2,2,6,6-Tetramethyl-tetrahydro-pyran-4-ol 95%
Dodavatel: Apollo Scientific
OR923261-1G
OR923261-250MG
APOSOR923261-1GEA
22950
CZK
APOSOR923261-1G
APOSOR923261-250MG
2,2,6,6-Tetramethyl-tetrahydro-pyran-4-ol 95%
2,2,6,6-Tetramethyl-tetrahydro-pyran-4-ol
Vzorec:
C₉H₁₈O₂ MW: 158,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21604057 CAS číslo: 20931-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...