Tisk…

3-Phenylcyclobutane-1-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Phenylcyclobutanecarboxylic acid

OR923203-1G OR923203-5G
APOSOR923203-1GEA 8300 CZK
APOSOR923203-1G APOSOR923203-5G
3-Phenylcyclobutane-1-carboxylic acid 97%
3-Phenylcyclobutane-1-carboxylic acid
Vzorec: C₁₁H₁₂O₂
MW: 176,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01734191
CAS číslo: 66016-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR