Tisk…

(3-Chloro-phenyl)-oxo-acetonitrile 95%

Dodavatel: Apollo Scientific

OR923195-1G OR923195-5G
APOSOR923195-1GEA 7250 CZK
APOSOR923195-1G APOSOR923195-5G
(3-Chloro-phenyl)-oxo-acetonitrile 95%
(3-Chloro-phenyl)-oxo-acetonitrile
Vzorec: C₈H₄ClNO
MW: 165,576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840530
CAS číslo: 26152-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR