Tisk…

Methyl-2-(benzylamino)acetate 97%

Dodavatel: Apollo Scientific

Synonyms: N-Benzylglycine methyl ester, Methyl 2-(benzylamino)acetate

OR923130-1G OR923130-25G
APOSOR923130-1GEA 870 CZK
APOSOR923130-1G APOSOR923130-25G
Methyl-2-(benzylamino)acetate 97%
Methyl-2-(benzylamino)acetate
Vzorec: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03247461
CAS číslo: 17136-35-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR