Tisk…

6-Methylpyrimidine-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR923073-1G OR923073-250MG
APOSOR923073-1GEA 7440 CZK
APOSOR923073-1G APOSOR923073-250MG
6-Methylpyrimidine-4-carboxylic acid 95%
6-Methylpyrimidine-4-carboxylic acid
Vzorec: C₆H₆N₂O₂
MW: 138,124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09881198
CAS číslo: 138538-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR