4-(2-Oxo-1-pyrrolidinyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
OR922930-1G
OR922930-250MG
APOSOR922930-1GEA
15940
CZK
APOSOR922930-1G
APOSOR922930-250MG
4-(2-Oxo-1-pyrrolidinyl)benzaldehyde 95%
4-(2-Oxo-1-pyrrolidinyl)benzaldehyde
Vzorec:
C₁₁H₁₁NO₂ MW: 189,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00454011 CAS číslo: 36151-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...