Tisk…

(S,E)-5-Chloro-2-isopropylpent-4-enoic acid 95%

Dodavatel: Apollo Scientific

OR922689-1G OR922689-250MG
APOSOR922689-1GEA 22950 CZK
APOSOR922689-1G APOSOR922689-250MG
(S,E)-5-Chloro-2-isopropylpent-4-enoic acid 95%
(S,E)-5-Chloro-2-isopropylpent-4-enoic acid
Vzorec: C₈H₁₃ClO₂
MW: 176,641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113211
CAS číslo: 324519-66-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR