Tisk…

Methyl 2-aminopentanoate hydrochloride 95%

Dodavatel: Apollo Scientific

OR922658-25G OR922658-5G
APOSOR922658-25GEA 9920 CZK
APOSOR922658-25G APOSOR922658-5G
Methyl 2-aminopentanoate hydrochloride 95%
Methyl 2-aminopentanoate hydrochloride
Vzorec: C₆H₁₃NO₂·ClH
MW: 167,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632063
CAS číslo: 10047-10-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR