6-Propylpyridin-2-amine 95%
Dodavatel: Apollo Scientific
OR922634-100MG
OR922634-1G
OR922634-250MG
APOSOR922634-100MGEA
2940
CZK
APOSOR922634-100MG
APOSOR922634-1G
APOSOR922634-250MG
6-Propylpyridin-2-amine 95%
6-Propylpyridin-2-amine
Vzorec:
C₈H₁₂N₂ MW: 136,194 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00130076 CAS číslo: 41995-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...