Methyl 6-aminopyrimidine-4-carboxylate 95%
Dodavatel: Apollo Scientific
OR922408-1G
OR922408-250MG
APOSOR922408-1GEA
22950
CZK
APOSOR922408-1G
APOSOR922408-250MG
Methyl 6-aminopyrimidine-4-carboxylate 95%
Methyl 6-aminopyrimidine-4-carboxylate
Vzorec:
C₆H₇N₃O₂ MW: 153,139 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13174840 CAS číslo: 77817-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...