2-Amino-2-methylpropane hydrochloride 95%
Dodavatel: Apollo Scientific
OR922309-1G
OR922309-5G
APOSOR922309-1GEA
612
CZK
APOSOR922309-1G
APOSOR922309-5G
2-Amino-2-methylpropane hydrochloride 95%
2-Amino-2-methylpropane hydrochloride
Vzorec:
ClH·C₄H₁₁N Storage Temperature: Ambient |
MDL Number:
MFCD00042027 CAS číslo: 10017-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...