Tisk…

2-Cyclohexyl-2-oxoacetic acid 95%

Dodavatel: Apollo Scientific

OR922240-1G OR922240-250MG
APOSOR922240-1GEA 22950 CZK
APOSOR922240-1G APOSOR922240-250MG
2-Cyclohexyl-2-oxoacetic acid 95%
2-Cyclohexyl-2-oxoacetic acid
Vzorec: C₈H₁₂O₃
MW: 156,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318468
CAS číslo: 4354-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR