1-[2-(Allyloxy)phenyl]ethanamine 95%
Dodavatel: Apollo Scientific
OR922195-1G
OR922195-250MG
APOSOR922195-1GEA
17990
CZK
APOSOR922195-1G
APOSOR922195-250MG
1-[2-(Allyloxy)phenyl]ethanamine 95%
1-[2-(Allyloxy)phenyl]ethanamine
Vzorec:
C₁₁H₁₅NO Storage Temperature: Ambient |
MDL Number:
MFCD12802686 CAS číslo: 1184578-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...