Tisk…

Boc-DL-Cha-OH

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-cyclohexylpropanoic acid

OR922005-10G OR922005-25G OR922005-5G
APOSOR922005-10GEA 5200 CZK
APOSOR922005-10G APOSOR922005-25G APOSOR922005-5G
Boc-DL-Cha-OH
Boc-DL-Cha-OH
Vzorec: C₁₄H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD03095514
CAS číslo: 144186-13-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR