(S)-2-Ethyl-piperazine 95%
Dodavatel: Apollo Scientific
Synonyms:
(2S)-2-Ethylpiperazine, (S)-2-Ethylpiperazine
OR921986-1G
OR921986-250MG
APOSOR921986-1GEA
22950
CZK
APOSOR921986-1G
APOSOR921986-250MG
(S)-2-Ethyl-piperazine 95%
(S)-2-Ethyl-piperazine
Vzorec:
C₆H₁₄N₂ MW: 114,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07373041 CAS číslo: 207284-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...