3-Chloro-6-methyl-1H-indazole 98%
Dodavatel: Apollo Scientific
OR921939-1G
OR921939-250MG
APOSOR921939-1GEA
17990
CZK
APOSOR921939-1G
APOSOR921939-250MG
3-Chloro-6-methyl-1H-indazole 98%
3-Chloro-6-methyl-1H-indazole
Vzorec:
C₈H₇ClN₂ MW: 166,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06738479 CAS číslo: 885271-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...