2-(2-Chlorophenyl)-2-methyl-1-propanol 95%
Dodavatel: Apollo Scientific
OR921792-1G
OR921792-250MG
APOSOR921792-1GEA
20900
CZK
APOSOR921792-1G
APOSOR921792-250MG
2-(2-Chlorophenyl)-2-methyl-1-propanol 95%
2-(2-Chlorophenyl)-2-methyl-1-propanol
Vzorec:
C₁₀H₁₃ClO MW: 184,663 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11916079 CAS číslo: 1176587-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...