Tisk…

1H-Pyrrolo[2,3-c]pyridine-3-carbonitrile 95%

Dodavatel: Apollo Scientific

OR921769-1G OR921769-250MG
APOSOR921769-1GEA 23970 CZK
APOSOR921769-1G APOSOR921769-250MG
1H-Pyrrolo[2,3-c]pyridine-3-carbonitrile 95%
1H-Pyrrolo[2,3-c]pyridine-3-carbonitrile
Vzorec: C₈H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD19221668
CAS číslo: 25957-69-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR