2-Aminomethyl-5-bromothiazole hydrochloride 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-Bromothiazol-2-yl)methanamine hydrochloride
OR921697-1G
OR921697-250MG
APOSOR921697-1GEA
22780
CZK
APOSOR921697-1G
APOSOR921697-250MG
2-Aminomethyl-5-bromothiazole hydrochloride 95+%
2-Aminomethyl-5-bromothiazole hydrochloride
Vzorec:
C₄H₅BrN₂S·ClH MW: 229,526 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22689362 CAS číslo: 1414958-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...