Tisk…

tert-Butyl 4-aminobutanoate 95%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 4-Aminobutanoate

OR921654-1G OR921654-250MG
APOSOR921654-1GEA 6750 CZK
APOSOR921654-1G APOSOR921654-250MG
tert-Butyl 4-aminobutanoate 95%
tert-Butyl 4-aminobutanoate
Vzorec: C₈H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD09923510
CAS číslo: 50479-22-6
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR